Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 7 - 8 | 1059 | 87 | |
| XM_021251571.1 | XP_021107230.1 | 7 - 8 | 1075 | 87 | ||
More Information
Set Membership: |
|

LOC101725179
Genomic Map