Assay Details
| Gene Symbol: LOC101725179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874014.2 | XP_004874071.1 | 8 - 9 | 1174 | 73 | |
| XM_021251571.1 | XP_021107230.1 | 8 - 9 | 1190 | 73 | ||
More Information
Set Membership: |
|

LOC101725179
Genomic Map