Assay Details
| Gene Symbol: Nudt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066154.2 | XP_012921608.1 | 3 - 4 | 6262 | 72 | |
| XM_013066164.2 | XP_012921618.1 | 3 - 4 | 6277 | 72 | ||
| XM_021243414.1 | XP_021099073.1 | 3 - 4 | 6260 | 72 | ||
More Information
Set Membership: |
|

Nudt14
Genomic Map