Assay Details
| Gene Symbol: Rarres1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071375.2 | XP_012926829.1 | 3 - 4 | 515 | 80 | |
| XM_013071380.2 | XP_012926834.1 | 3 - 4 | 515 | 80 | ||
More Information
Set Membership: |
|

Rarres1
Genomic Map