Assay Details
| Gene Symbol: LOC101726611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257772.1 | XP_021113431.1 | 10 - 11 | 1135 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726611| Gene Symbol: LOC101726611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257772.1 | XP_021113431.1 | 10 - 11 | 1135 | 87 | |
Set Membership: |
|