Assay Details
| Gene Symbol: Mindy4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259348.1 | XP_021115007.1 | 2 - 3 | 138 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mindy4b| Gene Symbol: Mindy4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259348.1 | XP_021115007.1 | 2 - 3 | 138 | 84 | |