Assay Details
| Gene Symbol: Mindy4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259348.1 | XP_021115007.1 | 3 - 4 | 302 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mindy4b| Gene Symbol: Mindy4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259348.1 | XP_021115007.1 | 3 - 4 | 302 | 120 | |
Set Membership: |
|