Assay Details
| Gene Symbol: Vsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837207.2 | XP_004837264.1 | 1 - 2 | 372 | 81 | |
| XM_013065915.2 | XP_012921369.1 | 1 - 2 | 372 | 81 | ||
More Information
Set Membership: |
|

Vsx2
Genomic Map