Assay Details
| Gene Symbol: Alk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839247.3 | XP_004839304.1 | 9 - 10 | 2309 | 58 | |
| XM_021253678.1 | XP_021109337.1 | 9 - 10 | 2313 | 58 | ||
| XM_021253679.1 | XP_021109338.1 | 10 - 11 | 1444 | 58 | ||
| XM_021253680.1 | XP_021109339.1 | 10 - 11 | 1334 | 58 | ||
More Information
Set Membership: |
|

Alk
Genomic Map