Assay Details
| Gene Symbol: Apcdd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841166.3 | XP_004841223.1 | 1 - 2 | 855 | 94 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apcdd1l| Gene Symbol: Apcdd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841166.3 | XP_004841223.1 | 1 - 2 | 855 | 94 | |