Assay Details
| Gene Symbol: Spg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842640.3 | XP_004842697.1 | 3 - 4 | 421 | 58 | |
| XM_004842641.3 | XP_004842698.1 | 3 - 4 | 414 | 58 | ||
| XM_004842642.3 | XP_004842699.1 | 3 - 4 | 422 | 58 | ||
More Information
Set Membership: |
|

Spg7
Genomic Map