Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845599.2 | XP_004845656.1 | 4 - 5 | 451 | 56 | |
| XM_004845600.2 | XP_004845657.1 | 4 - 5 | 451 | 56 | ||
| XM_004845601.3 | XP_004845658.1 | 4 - 5 | 451 | 56 | ||
More Information
Set Membership: |
|

Cby1
Genomic Map