Assay Details
| Gene Symbol: Med30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850201.3 | XP_004850258.1 | 3 - 4 | 560 | 102 | |
| XM_021262719.1 | XP_021118378.1 | 3 - 4 | 561 | 102 | ||
More Information
Set Membership: |
|

Med30
Genomic Map