Assay Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853147.2 | XP_004853204.1 | 4 - 5 | 1317 | 63 | |
| XM_021265146.1 | XP_021120805.1 | 4 - 5 | 1220 | 63 | ||
| XM_021265147.1 | XP_021120806.1 | 4 - 5 | 1206 | 63 | ||
More Information
Set Membership: |
|

Cabyr
Genomic Map