Assay Details
| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854776.3 | XP_004854833.1 | 13 - 14 | 1200 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3glct| Gene Symbol: B3glct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854776.3 | XP_004854833.1 | 13 - 14 | 1200 | 64 | |
Set Membership: |
|