Assay Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856339.3 | XP_004856396.1 | 6 - 7 | 839 | 66 | |
| XM_021237538.1 | XP_021093197.1 | 4 - 5 | 749 | 66 | ||
More Information
Set Membership: |
|

Ube2h
Genomic Map