Assay Details
| Gene Symbol: LOC101707284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859394.2 | XP_004859451.1 | 7 - 8 | 1326 | 81 | |
| XM_021240146.1 | XP_021095805.1 | 7 - 8 | 1404 | 81 | ||
More Information
Set Membership: |
|

LOC101707284
Genomic Map