Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860949.3 | XP_004861006.1 | 10 - 11 | 4380 | 63 | |
| XM_004860950.2 | XP_004861007.1 | 11 - 12 | 1506 | 63 | ||
| XM_013078199.2 | XP_012933653.1 | 10 - 11 | 1617 | 63 | ||
| XM_013078200.2 | XP_012933654.1 | 10 - 11 | 1648 | 63 | ||
| XM_021241130.1 | XP_021096789.1 | 9 - 10 | 4497 | 63 | ||
More Information
Set Membership: |
|

Gcgr
Genomic Map