Assay Details
| Gene Symbol: LOC101703228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865960.3 | XP_004866017.1 | 12 - 13 | 2504 | 62 | |
| XM_013065931.2 | XP_012921385.1 | 12 - 13 | 2505 | 62 | ||
| XM_013065933.2 | XP_012921387.1 | 12 - 13 | 2038 | 62 | ||
| XM_021245171.1 | XP_021100830.1 | 12 - 13 | 2505 | 62 | ||
| XM_021245172.1 | XP_021100831.1 | 11 - 12 | 2455 | 62 | ||
| XM_021245173.1 | XP_021100832.1 | 11 - 12 | 1933 | 62 | ||
More Information
Set Membership: |
|

LOC101703228
Genomic Map