Assay Details
| Gene Symbol: LOC101703228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865960.3 | XP_004866017.1 | 13 - 14 | 2586 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101703228| Gene Symbol: LOC101703228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865960.3 | XP_004866017.1 | 13 - 14 | 2586 | 86 | |
Set Membership: |
|