Assay Details
| Gene Symbol: Crocc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247330.1 | XP_021102989.1 | 22 - 23 | 3597 | 56 | |
| XM_021247331.1 | XP_021102990.1 | 22 - 23 | 3598 | 56 | ||
More Information
Set Membership: |
|

Crocc2
Genomic Map