Assay Details
| Gene Symbol: LOC101707839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248860.1 | XP_021104519.1 | 5 - 6 | 1071 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707839| Gene Symbol: LOC101707839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248860.1 | XP_021104519.1 | 5 - 6 | 1071 | 61 | |
Set Membership: |
|