Assay Details
Gene Symbol: LOC101702210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250026.1 | XP_021105685.1 | 3 - 4 | 345 | 81 | |
XM_021250027.1 | XP_021105686.1 | 3 - 4 | 241 | 81 |
