Assay Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251107.1 | XP_021106766.1 | 5 - 6 | 4259 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsh2d| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251107.1 | XP_021106766.1 | 5 - 6 | 4259 | 63 | |
Set Membership: |
|