Assay Details
| Gene Symbol: Capn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253690.1 | XP_021109349.1 | 19 - 20 | 2055 | 58 | |
| XM_021253691.1 | XP_021109350.1 | 18 - 19 | 1909 | 58 | ||
| XM_021253692.1 | XP_021109351.1 | 19 - 20 | 2552 | 58 | ||
More Information
Set Membership: |
|

Capn14
Genomic Map