Assay Details
| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843673.2 | XP_004843730.1 | 1 - 2 | 643 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004843673.2 transcripts.
Set Membership: |
|

Pla2g1b
Genomic Map