Assay Details
| Gene Symbol: LOC101710080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849462.1 | XP_004849519.1 | 13 - 14 | 1739 | 108 | |
| XM_013073872.1 | XP_012929326.1 | 13 - 14 | 1712 | 108 | ||
More Information
Set Membership: |
|

LOC101710080
Genomic Map