Assay Details
| Gene Symbol: Mindy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856284.2 | XP_004856341.1 | 4 - 5 | 772 | 58 | |
| XM_004856285.2 | XP_004856342.1 | 4 - 5 | 772 | 58 | ||
| XM_004856286.2 | XP_004856343.1 | 4 - 5 | 772 | 58 | ||
| XM_004856287.2 | XP_004856344.1 | 4 - 5 | 728 | 58 | ||
| XM_021237565.1 | XP_021093224.1 | 4 - 5 | 772 | 58 | ||
| XM_021237566.1 | XP_021093225.1 | 4 - 5 | 772 | 58 | ||
| XM_021237567.1 | XP_021093226.1 | 4 - 5 | 772 | 58 | ||
| XM_021237568.1 | XP_021093227.1 | 4 - 5 | 772 | 58 | ||
| XM_021237569.1 | XP_021093228.1 | 4 - 5 | 772 | 58 | ||
More Information
Set Membership: |
|

Mindy4
Genomic Map