Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861617.3 | XP_004861674.1 | 6 - 7 | 803 | 65 | |
| XM_004861620.2 | XP_004861677.1 | 6 - 7 | 787 | 65 | ||
| XM_021241827.1 | XP_021097486.1 | 5 - 6 | 643 | 65 | ||
| XM_021241828.1 | XP_021097487.1 | 3 - 4 | 358 | 65 | ||
| XM_021241829.1 | XP_021097488.1 | 4 - 5 | 471 | 65 | ||
More Information
Set Membership: |
|

Tyro3
Genomic Map