Assay Details
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863114.3 | XP_004863171.1 | 4 - 5 | 435 | 60 | |
| XM_013079077.2 | XP_012934531.1 | 5 - 6 | 559 | 60 | ||
| XM_013079078.2 | XP_012934532.2 | 3 - 4 | 421 | 60 | ||
| XM_013079079.2 | XP_012934533.2 | 3 - 4 | 421 | 60 | ||
| XM_021243013.1 | XP_021098672.1 | 6 - 7 | 1416 | 60 | ||
| XM_021243014.1 | XP_021098673.1 | 6 - 7 | 1275 | 60 | ||
| XM_021243015.1 | XP_021098674.1 | 6 - 7 | 1404 | 60 | ||
| XM_021243016.1 | XP_021098675.1 | 5 - 6 | 1327 | 60 | ||
| XM_021243017.1 | XP_021098676.1 | 6 - 7 | 1263 | 60 | ||
| XM_021243018.1 | XP_021098677.1 | 4 - 5 | 480 | 60 | ||
More Information
Set Membership: |
|

Vdr
Genomic Map