Assay Details
| Gene Symbol: Npm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866367.2 | XP_004866424.1 | 5 - 6 | 564 | 82 | |
| XM_013066086.2 | XP_012921540.1 | 5 - 6 | 741 | 82 | ||
| XM_013066087.2 | XP_012921541.1 | 5 - 6 | 714 | 82 | ||
| XM_013066089.2 | XP_012921543.1 | 5 - 6 | 537 | 82 | ||
More Information
Set Membership: |
|

Npm3
Genomic Map