Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873219.3 | XP_004873276.1 | 6 - 7 | 1067 | 65 | |
| XM_013068674.2 | XP_012924128.1 | 6 - 7 | 1269 | 65 | ||
| XM_013068675.2 | XP_012924129.1 | 6 - 7 | 1034 | 65 | ||
| XM_013068676.2 | XP_012924130.1 | 6 - 7 | 1015 | 65 | ||
| XM_013068677.2 | XP_012924131.1 | 6 - 7 | 971 | 65 | ||
| XM_021251079.1 | XP_021106738.1 | 6 - 7 | 1068 | 65 | ||
More Information
Set Membership: |
|

Slc27a1
Genomic Map