Assay Details
| Gene Symbol: LOC101698345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066241.2 | XP_012921695.1 | 6 - 7 | 1427 | 68 | |
| XM_013066242.2 | XP_012921696.1 | 5 - 6 | 1288 | 68 | ||
| XM_021246093.1 | XP_021101752.1 | 5 - 6 | 1267 | 68 | ||
More Information
Set Membership: |
|

LOC101698345
Genomic Map