Assay Details
| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078075.2 | XP_012933529.1 | 3 - 4 | 1084 | 69 | |
| XM_021240863.1 | XP_021096522.1 | 3 - 4 | 543 | 69 | ||
More Information
Set Membership: |
|

Galntl5
Genomic Map