Assay Details
| Gene Symbol: LOC101698672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238054.1 | XP_021093713.1 | 26 - 27 | 3859 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698672| Gene Symbol: LOC101698672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238054.1 | XP_021093713.1 | 26 - 27 | 3859 | 88 | |
Set Membership: |
|