Assay Details
| Gene Symbol: LOC101711927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251200.1 | XP_021106859.1 | 1 - 2 | 3029 | 88 | |
| XM_021251202.1 | XP_021106861.1 | 1 - 2 | 3025 | 88 | ||
| XM_021251203.1 | XP_021106862.1 | 1 - 2 | 67 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021251203.1 transcripts.
Set Membership: |
|

LOC101711927
Genomic Map