Assay Details
| Gene Symbol: LOC101698709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256824.1 | XP_021112483.1 | 2 - 3 | 613 | 66 | |
| XM_021256825.1 | XP_021112484.1 | 2 - 3 | 613 | 66 | ||
More Information
Set Membership: |
|

LOC101698709
Genomic Map