Assay Details
| Gene Symbol: LOC101698709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256824.1 | XP_021112483.1 | 5 - 6 | 956 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698709| Gene Symbol: LOC101698709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256824.1 | XP_021112483.1 | 5 - 6 | 956 | 100 | |
Set Membership: |
|