Assay Details
| Gene Symbol: Washc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258308.1 | XP_021113967.1 | 30 - 31 | 3172 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Washc4| Gene Symbol: Washc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258308.1 | XP_021113967.1 | 30 - 31 | 3172 | 67 | |
Set Membership: |
|