Assay Details
| Gene Symbol: B4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838395.3 | XP_004838452.1 | 2 - 3 | 753 | 74 | |
| XM_004838396.3 | XP_004838453.1 | 2 - 3 | 756 | 74 | ||
More Information
Set Membership: |
|

B4galt1
Genomic Map