Assay Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839928.2 | XP_004839985.1 | 9 - 10 | 1398 | 47 | |
| XM_013070443.2 | XP_012925897.1 | 8 - 9 | 1288 | 47 | ||
| XM_021254287.1 | XP_021109946.1 | 9 - 10 | 1461 | 47 | ||
| XM_021254288.1 | XP_021109947.1 | 8 - 9 | 1363 | 47 | ||
More Information
Set Membership: |
|

Polm
Genomic Map