Assay Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845755.3 | XP_004845812.1 | 3 - 4 | 837 | 94 | |
| XM_021259102.1 | XP_021114761.1 | 2 - 3 | 184 | 94 | ||
More Information
Set Membership: |
|

Etnk1
Genomic Map