Assay Details
| Gene Symbol: LOC101718101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847589.3 | XP_004847646.1 | 12 - 13 | 1721 | 94 | |
| XM_021260544.1 | XP_021116203.1 | 12 - 13 | 1721 | 94 | ||
More Information
Set Membership: |
|

LOC101718101
Genomic Map