Assay Details
| Gene Symbol: LOC101718101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847589.3 | XP_004847646.1 | 16 - 17 | 2282 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101718101| Gene Symbol: LOC101718101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847589.3 | XP_004847646.1 | 16 - 17 | 2282 | 92 | |
Set Membership: |
|