Assay Details
| Gene Symbol: LOC101718101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847589.3 | XP_004847646.1 | 3 - 4 | 527 | 83 | |
| XM_021260544.1 | XP_021116203.1 | 3 - 4 | 527 | 83 | ||
More Information
Set Membership: |
|

LOC101718101
Genomic Map