Assay Details
| Gene Symbol: Naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848048.2 | XP_004848105.1 | 2 - 3 | 400 | 66 | |
| XM_004848049.2 | XP_004848106.1 | 2 - 3 | 271 | 66 | ||
| XM_021261050.1 | XP_021116709.1 | 2 - 3 | 400 | 66 | ||
More Information
Set Membership: |
|

Naaa
Genomic Map