Assay Details
| Gene Symbol: LOC101722877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848053.2 | XP_004848110.1 | 6 - 7 | 1377 | 81 | |
| XM_021261056.1 | XP_021116715.1 | 7 - 8 | 1414 | 81 | ||
More Information
Set Membership: |
|

LOC101722877
Genomic Map