Assay Details
Gene Symbol: LOC101722077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850388.2 | XP_004850445.1 | 15 - 16 | 1855 | 80 | |
XM_021262904.1 | XP_021118563.1 | 16 - 17 | 1993 | 80 | ||
XM_021262905.1 | XP_021118564.1 | 16 - 17 | 1990 | 80 | ||
XM_021262906.1 | XP_021118565.1 | 16 - 17 | 1972 | 80 | ||
XM_021262907.1 | XP_021118566.1 | 15 - 16 | 1876 | 80 | ||
XM_021262908.1 | XP_021118567.1 | 14 - 15 | 1722 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
