Assay Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854713.2 | XP_004854770.1 | 2 - 3 | 696 | 62 | |
| XM_021266433.1 | XP_021122092.1 | 3 - 4 | 762 | 62 | ||
More Information
Set Membership: |
|

Abcb10
Genomic Map