Assay Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857308.3 | XP_004857365.1 | 3 - 4 | 657 | 71 | |
| XM_004857309.3 | XP_004857366.1 | 3 - 4 | 482 | 71 | ||
| XM_004857310.3 | XP_004857367.1 | 3 - 4 | 625 | 71 | ||
More Information
Set Membership: |
|

Eno3
Genomic Map